Anti-cry1Ab Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A51695-100UG A51695-50UG
77817-661EA 330 USD
77817-661 77817-662
Anti-cry1Ab Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: cry1Ab
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacillus thuringiensis subsp. kurstaki
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR