Anti-lepB Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A51539-100UG A51539-50UG
77817-363EA 330 USD
77817-363 77817-364
Anti-lepB Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: lepB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR