Anti-crp Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A51518-100UG A51518-50UG
77817-324EA 330 USD
77817-324 77817-325
Anti-crp Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: crp
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR