Anti-HLA-C Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A51299-50UG A51299-100UG
77816-916EA 225 USD
77816-916 77816-915
Anti-HLA-C Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: HLA-C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR