Anti-eno Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A51210-100UG A51210-50UG
77816-745EA 330 USD
77816-745 77816-746
Anti-eno Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: eno
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Staphylococcus aureus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR