Anti-TES-26 Rabbit Recombinant Antibody

Supplier: AFG Bioscience
A51137-100UG A51137-50UG
77816-607EA 330 USD
77816-607 77816-608
Anti-TES-26 Rabbit Recombinant Antibody
Antibodies
Type:
Antigen: TES-26
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Toxocara canis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR