Anti-AFP2 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A50936-100UG A50936-50UG
77816-228EA 330 USD
77816-228 77816-229
Anti-AFP2 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: AFP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Raphanus sativus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR