Anti-ompA Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))

Supplier: AFG Bioscience
A50801-50UG A50801-100UG
77815-976EA 225 USD
77815-976 77815-975
Anti-ompA Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type:
Antigen: ompA
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chlamydophila pneumoniae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR