Anti-N Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A50315-50UG
77815-053EA 225 USD
77815-053
Anti-N Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:5000

Type:
Antigen: N
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Junin mammarenavirus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR