Anti-ACY1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49711-100UL A49711-50UL
77813-977EA 330 USD
77813-977 77813-978
Anti-ACY1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100

Type:
Antigen: ACY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR