Anti-PAPPA Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49467-100UL A49467-50UL
77813-561EA 330 USD
77813-561 77813-562
Anti-PAPPA Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200

Type:
Antigen: PAPPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR