Anti-FGFR1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49355-100UL
77813-380EA 330 USD
77813-380
Anti-FGFR1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:3000, IHC: 1:50 to 1:100

Type:
Antigen: FGFR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR