Anti-RNF148 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49195-100UL A49195-50UL
77813-121EA 330 USD
77813-121 77813-122
Anti-RNF148 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200

Type:
Antigen: RNF148
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR