Anti-CD97 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43009-50UG
77812-439EA 150 USD
77812-439
Anti-CD97 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500 to 1:2000, ELISA:1:5000

Type:
Antigen: CD97
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR