Anti-CALCRL Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A42946-100UL A42946-50UL
77812-373EA 330 USD
77812-373 77812-374
Anti-CALCRL Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA:1:1000 to 1:2000, WB: 1:200 to 1:1000

Type:
Antigen: CALCRL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR