Anti-Csf1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A42815-100UG A42815-50UG
77812-154EA 330 USD
77812-154 77812-155
Anti-Csf1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: Csf1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR