Anti-UCN Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A37985-100UL
77811-932EA 585 USD
77811-932
Anti-UCN Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: UCN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR