Anti-UBE2D3 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A37869-50UG
77811-841EA 225 USD
77811-841
Anti-UBE2D3 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500 to 1:5000

Type:
Antigen: UBE2D3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR