Anti-UBA52 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A37823-100UG A37823-50UG
77811-803EA 330 USD
77811-803 77803-803
Anti-UBA52 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: UBA52
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR