Anti-TYK2 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A37779-100UG A37779-50UG
77811-767EA 330 USD
77811-767 77803-767
Anti-TYK2 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200

Type:
Antigen: TYK2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR