Anti-Ttr Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A37676-100UG A37676-50UG
77811-683EA 330 USD
77811-683 77803-683
Anti-Ttr Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: Ttr
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR