Anti-RPL17 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A37550-100UG A37550-50UG
77811-584EA 330 USD
77811-584 77803-584
Anti-RPL17 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:5000

Type:
Antigen: RPL17
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR