Anti-TRIM37 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A37501-100UG A37501-50UG
77811-547EA 330 USD
77811-547 77803-547
Anti-TRIM37 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: TRIM37
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR