Anti-TPTE Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A37367-100UG A37367-50UG
77811-435EA 330 USD
77811-435 77803-435
Anti-TPTE Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: TPTE
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR