Anti-TPST1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A37346-100UG A37346-50UG
77811-416EA 330 USD
77811-416 77803-416
Anti-TPST1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: TPST1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR