Anti-Tpsb2 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A37340-100UG A37340-50UG
77811-411EA 330 USD
77811-411 77803-411
Anti-Tpsb2 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: Tpsb2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR