Anti-TPD52L2 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A37279-100UG A37279-50UG
77811-361EA 330 USD
77811-361 77803-361
Anti-TPD52L2 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: TPD52L2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR