Anti-TP73 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A37270-100UL A37270-50UL
77811-354EA 330 USD
77811-354 77803-354
Anti-TP73 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: TP73
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR