Anti-TOR1A Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A37241-100UG A37241-50UG
77811-329EA 330 USD
77811-329 77803-329
Anti-TOR1A Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: TOR1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR