Anti-TOMM40 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A37220-100UL A37220-50UL
77811-313EA 330 USD
77811-313 77803-313
Anti-TOMM40 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: TOMM40
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR