Anti-TNFSF12 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A37140-100UG A37140-50UG
77811-249EA 330 USD
77811-249 77803-249
Anti-TNFSF12 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: TNFSF12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR