Anti-Tnfsf11 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A37133-100UG A37133-50UG
77811-242EA 330 USD
77811-242 77803-242
Anti-Tnfsf11 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: Tnfsf11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR