Anti-TMEM27 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A36906-100UL
77811-037EA 585 USD
77811-037
Anti-TMEM27 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TMEM27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR