Anti-RAB11FIP1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A41589-50UG
77809-288EA 225 USD
77809-288
Anti-RAB11FIP1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:20000

Type:
Antigen: RAB11FIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR