Anti-LYPLA2 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A40991-100UG A40991-50UG
77808-368EA 330 USD
77808-368 77808-369
Anti-LYPLA2 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IF: 1:50 to 1:200

Type:
Antigen: LYPLA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR