Anti-TGFBR2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A40911-50UG
77808-274EA 225 USD
77808-274
Anti-TGFBR2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:10000

Type:
Antigen: TGFBR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR