Anti-TBXAS1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A40880-50UG
77808-243EA 225 USD
77808-243
Anti-TBXAS1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:20000

Type:
Antigen: TBXAS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR