Anti-MAP3K7 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A40874-50UG
77808-237EA 225 USD
77808-237
Anti-MAP3K7 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:20000

Type:
Antigen: MAP3K7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR