Anti-BRS3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A40846-50UG
77808-209EA 225 USD
77808-209
Anti-BRS3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA:1:10000

Type:
Antigen: BRS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR