Anti-ANAPC1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A40722-50UG
77808-081EA 225 USD
77808-081
Anti-ANAPC1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA:1:10000

Type:
Antigen: ANAPC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR