Anti-CASP12 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A40377-100UL A40377-50UL
77807-525EA 330 USD
77807-525 77807-526
Anti-CASP12 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:10000, IHC: 1:50 to 1:200

Type:
Antigen: CASP12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR