Anti-IFT74 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A40347-100UL A40347-50UL
77807-471EA 330 USD
77807-471 77807-472
Anti-IFT74 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:50 to 1:200

Type:
Antigen: IFT74
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR