Anti-GFRA3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A39957-100UL A39957-50UL
77807-008EA 330 USD
77807-008 77807-009
Anti-GFRA3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:5000, WB: 1:1000 to 1:3000, IHC: 1:20 to 1:100

Type:
Antigen: GFRA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR