Anti-TMPRSS5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A39901-100UL A39901-50UL
77806-907EA 330 USD
77806-907 77806-908
Anti-TMPRSS5 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:10 to 1:50

Type:
Antigen: TMPRSS5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR