Anti-ZNF592 Rabbit Recombinant Antibody

Supplier: AFG Bioscience
A39897-50UG A39897-100UG
77806-900EA 225 USD
77806-900 77806-899
Anti-ZNF592 Rabbit Recombinant Antibody
Antibodies
Type:
Antigen: ZNF592
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR