Anti-UBA52 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A39862-100UL
77806-841EA 330 USD
77806-841
Anti-UBA52 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:50 to 1:100

Type:
Antigen: UBA52
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR