Anti-DPYSL4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A39818-100UL A39818-50UL
77806-760EA 330 USD
77806-760 77806-761
Anti-DPYSL4 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:500 to 1:5000, WB: 1:500 to 1:2000

Type:
Antigen: DPYSL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR