Anti-EIF3I Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A39675-100UG A39675-50UG
77806-498EA 330 USD
77806-498 77806-499
Anti-EIF3I Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: EIF3I
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR