Anti-AGFG1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A39654-100UL A39654-50UL
77806-468EA 330 USD
77806-468 77806-469
Anti-AGFG1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200

Type:
Antigen: AGFG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR