Anti-CDK5RAP2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A39550-50UG
77806-344EA 225 USD
77806-344
Anti-CDK5RAP2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:20000

Type:
Antigen: CDK5RAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR